accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH3CNHOOONHSOOCH3

CAS RN 107753-78-6

IUPAC Name: cyclopentyl N-[3-({2-methoxy-4-[(2-methylbenzenesulfonyl)carbamoyl]phenyl}methyl)-1-methyl-1H-indol-5-yl]carbamate
Synonyms: zafirlukast aeronix menarini brand of zafirlukast zafirlukast (product) novartis brand of zafirlukast accolate astrazeneca brand of zafirlukast zafirlukast (substance) zafirlukast preparation zeneca brand of zafirkulast
Molecular Weight (g/mol): 575.68
Molecular Formula: C31H33N3O6S
InChi Key: YEEZWCHGZNKEEK-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1CC1=CN(C)C2=CC=C(NC(=O)OC3CCCC3)C=C12)C(=O)NS(=O)(=O)C1=CC=CC=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results