accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONONOO

CAS RN 1078-05-3

IUPAC Name: 3-carboxy-4-nitropyridin-1-ium-1-olate
Synonyms: 3-carboxy-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 184.11
Molecular Formula: C6H4N2O5
InChi Key: OYLXIYDCLHUDQV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C[N+]([O-])=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results