accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1078-19-9

IUPAC Name: 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 6-methoxy-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: MNALUTYMBUBKNX-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(=O)CCCC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (4)
  • (5)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)