accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1078-61-1

IUPAC Name: 3-(3,4-dihydroxyphenyl)propanoic acid
Synonyms: dihydrocaffeic acid hydrocaffeate hydrocaffeic acid 3,4-dihydroxydihydrocinnamate dihydrocaffeate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: DZAUWHJDUNRCTF-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)