accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNNOOH3CH3CCH3SH3C(E/Z)

CAS RN 107819-90-9

IUPAC Name: tert-butyl N-({[(tert-butoxy)carbonyl]amino}(methylsulfanyl)methylidene)carbamate
Synonyms: tert-butyl N-{[(tert-butoxycarbonyl)amino](methylsulfanyl)methylidene}carbamate
Molecular Weight (g/mol): 290.38
Molecular Formula: C12H22N2O4S
InChi Key: UQJXXWHAJKRDKY-UHFFFAOYSA-N
SMILES: CSC(NC(=O)OC(C)(C)C)=NC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)