accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOHO

CAS RN 107873-03-0

IUPAC Name: 2,2-difluorocyclopropane-1-carboxylic acid
Synonyms: 2,2-difluorocyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 122.07
Molecular Formula: C4H4F2O2
InChi Key: KMLMOVWSQPHQME-UHFFFAOYNA-N
SMILES: OC(=O)C1CC1(F)F
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)