accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNHH2N

CAS RN 107890-32-4

IUPAC Name: 4-{[4-(trifluoromethyl)phenyl]methyl}piperazine-1,4-diium
Synonyms: 4-{[4-(trifluoromethyl)phenyl]methyl}piperazine-1,4-diium
Molecular Weight (g/mol): 246.28
Molecular Formula: C12H17F3N2
InChi Key: FAFAFWFQFVLXGF-UHFFFAOYSA-P
SMILES: FC(F)(F)C1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results