accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1079-17-0

IUPAC Name: 1,2,4,5-tetrachloro-3,6-bis(chloromethyl)benzene
Synonyms: 1,2,4,5-tetrachloro-3,6-bis(chloromethyl)benzene
Molecular Weight (g/mol): 312.82
Molecular Formula: C8H4Cl6
InChi Key: IYGDLOMSJZQSGY-UHFFFAOYSA-N
SMILES: ClCC1=C(Cl)C(Cl)=C(CCl)C(Cl)=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results