accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1079054-78-6

IUPAC Name: 5-amino-6-methylpyridine-2-carbonitrile
Synonyms: 5-amino-6-methylpyridine-2-carbonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: SLBIKHREAIGBFI-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(=N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results