accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1079129-48-8

IUPAC Name: bis(fluorosulfonyl)azanide; diethyl(2-methoxyethyl)methylazanium
Synonyms: bis(fluorosulfonyl)azanide; diethyl(2-methoxyethyl)methylazanium
Molecular Weight (g/mol): 326.37
Molecular Formula: C8H20F2N2O5S2
InChi Key: OHKPLVVWXQGNTL-UHFFFAOYSA-N
SMILES: FS(=O)(=O)[N-]S(F)(=O)=O.CC[N+](C)(CC)CCOC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results