accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3H2N

CAS RN 108-00-9

IUPAC Name: (2-aminoethyl)dimethylamine
Synonyms: (2-aminoethyl)dimethylamine
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: DILRJUIACXKSQE-UHFFFAOYSA-N
SMILES: CN(C)CCN
Boiling Point 
  • (4)
  • (3)
  • (6)
Molecular Weight (g/mol) 
  • (7)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (3)
  • (6)
  • (4)
  • (3)
  • (6)
  • (7)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)