accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-09-8

CH3H3CCH3H2N

CAS RN 108-09-8

IUPAC Name: 4-methylpentan-2-amine
Synonyms: 2-pentanamine, 4-methyl-
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: UNBMPKNTYKDYCG-UHFFFAOYNA-N
SMILES: CC(C)CC(C)N
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)