accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-26-9

IUPAC Name: 3-methyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 3-methyl-5-pyrazolone
Molecular Weight (g/mol): 98.11
Molecular Formula: C4H6N2O
InChi Key: NHLAPJMCARJFOG-UHFFFAOYSA-N
SMILES: CC1=NNC(=O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results