accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-27-0

IUPAC Name: 5-methylpyrrolidin-2-one
Synonyms: 2-pyrrolidinone, 5-methyl-
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: YVIVRJLWYJGJTJ-UHFFFAOYNA-N
SMILES: CC1CCC(=O)N1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)