accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-30-5

IUPAC Name: oxolane-2,5-dione
Synonyms: succinic anhydride rikacid SA bernsteinsaeureanhydrid bernsteinsaure-anhydrid succinyl anhydride
Molecular Weight (g/mol): 100.07
Molecular Formula: C4H4O3
InChi Key: RINCXYDBBGOEEQ-UHFFFAOYSA-N
SMILES: O=C1CCC(=O)O1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (1)
  • (2)
  • (2)
  • (9)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (9)
  • (2)