accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-33-8

CH3SNH2NN

CAS RN 108-33-8

IUPAC Name: 5-methyl-1,3,4-thiadiazol-2-amine
Synonyms: 5-methyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 115.15
Molecular Formula: C3H5N3S
InChi Key: HMPUHXCGUHDVBI-UHFFFAOYSA-N
SMILES: CC1=NN=C(N)S1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)