accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2NN

CAS RN 108-52-1

IUPAC Name: 4-methylpyrimidin-2-amine
Synonyms: 2-amino-4-methylpyrimidine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: GHCFWKFREBNSPC-UHFFFAOYSA-N
SMILES: CC1=CC=NC(N)=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)