accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-56-5

IUPAC Name: 1,4-diethyl 2-oxobutanedioate
Synonyms: ethyl oxalacetate
Molecular Weight (g/mol): 188.18
Molecular Formula: C8H12O5
InChi Key: JDXYSCUOABNLIR-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results