accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H2N

CAS RN 108-69-0

IUPAC Name: 3,5-dimethylaniline
Synonyms: 3,5-dimethylaniline
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: MKARNSWMMBGSHX-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC(C)=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)