accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-74-7

IUPAC Name: 1,3,5-trimethyl-1,3,5-triazinane
Synonyms: 1,3,5-trimethyl-1,3,5-triazinane
Molecular Weight (g/mol): 129.21
Molecular Formula: C6H15N3
InChi Key: DPMZXMBOYHBELT-UHFFFAOYSA-N
SMILES: CN1CN(C)CN(C)C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)