accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHCH3CH3

CAS RN 108-82-7

IUPAC Name: 2,6-dimethylheptan-4-ol
Synonyms: 2,6-dimethyl-4-heptanol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: HXQPUEQDBSPXTE-UHFFFAOYSA-N
SMILES: CC(C)CC(O)CC(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)