accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-91-8

IUPAC Name: cyclohexanamine
Synonyms: cyclohexylamine carbonic acid, compd. with cyclohexanamine (9CI)
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: PAFZNILMFXTMIY-UHFFFAOYSA-N
SMILES: NC1CCCCC1
Boiling Point 
  • (3)
  • (4)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)