accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 108-97-4

IUPAC Name: 4H-pyran-4-one
Synonyms: pyran-4-one gamma-pyrone
Molecular Weight (g/mol): 96.09
Molecular Formula: C5H4O2
InChi Key: CVQUWLDCFXOXEN-UHFFFAOYSA-N
SMILES: O=C1C=COC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)