accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108-99-6

IUPAC Name: 3-methylpyridine
Synonyms: 3-methylpyridine 3-mepy
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: ITQTTZVARXURQS-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1
Boiling Point 
  • (2)
  • (2)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (8)
Percent Purity 
  • (2)
  • (1)
  • (9)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (8)
  • (2)
  • (1)
  • (9)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)