accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 1080-74-6

IUPAC Name: 2-(3-oxo-2,3-dihydro-1H-inden-1-ylidene)propanedinitrile
Synonyms: 2-(3-oxo-2H-inden-1-ylidene)propanedinitrile
Molecular Weight (g/mol): 194.19
Molecular Formula: C12H6N2O
InChi Key: QNVKZKOSAXYVFZ-UHFFFAOYSA-N
SMILES: O=C1CC(=C(C#N)C#N)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results