accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108002-87-5

IUPAC Name: 2-(phenylamino)pyrimidine-5-carbaldehyde
Synonyms: 2-(phenylamino)pyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 199.21
Molecular Formula: C11H9N3O
InChi Key: QNFHXIZSGIGYOU-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(NC2=CC=CC=C2)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results