accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1080028-74-5

IUPAC Name: tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
Molecular Weight (g/mol): 319.4
Molecular Formula: C18H25NO4
InChi Key: WQFHJESQGSVMDW-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)OCC1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results