accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108128-39-8

IUPAC Name: 5-(furan-2-yl)-1-methyl-1H-pyrazole-3-carboxylic acid
Synonyms: 5-(furan-2-yl)-1-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C9H8N2O3
InChi Key: XXEWHNSDJMBLBK-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CO1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results