accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)OH3CCH3NOOCH3CH3H3C

CAS RN 108149-60-6

IUPAC Name: 3-tert-butyl 4-methyl (4S)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
Synonyms: 3-tert-butyl 4-methyl (4S)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
Molecular Weight (g/mol): 259.3
Molecular Formula: C12H21NO5
InChi Key: ZNBUXTFASGDVCL-QMMMGPOBSA-N
SMILES: COC(=O)[C@@H]1COC(C)(C)N1C(=O)OC(C)(C)C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results