accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108166-02-5

IUPAC Name: (2-methylquinolin-6-yl)methanol
Synonyms: (2-methylquinolin-6-yl)methanol
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: HLFYJILNIBADGG-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(CO)C=CC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results