accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHH2NHClHCl

CAS RN 1082692-96-3

IUPAC Name: 2-amino-3-(pyridin-2-yl)propanoic acid dihydrochloride
Synonyms: 2-amino-3-(pyridin-2-yl)propanoic acid dihydrochloride
Molecular Weight (g/mol): 239.1
Molecular Formula: C8H12Cl2N2O2
InChi Key: HGOJCMDWQOEDJA-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(CC1=CC=CC=N1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)