accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOClNH2

CAS RN 108282-38-8

IUPAC Name: 4-amino-5-chloro-2-ethoxybenzoic acid
Synonyms: 4-amino-5-chloro-2-ethoxybenzoic acid
Molecular Weight (g/mol): 215.63
Molecular Formula: C9H10ClNO3
InChi Key: XWGYOMHQGQZRLC-UHFFFAOYSA-N
SMILES: CCOC1=CC(N)=C(Cl)C=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results