CAS RN 1083-41-6
CAS RN 1083-41-6
IUPAC Name:
butyl 3,4,5-trihydroxybenzoate
Synonyms:
butyl gallate
Molecular Weight (g/mol):
226.23
Molecular Formula:
C11H14O5
InChi Key:
XOPOEBVTQYAOSV-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
1
–
1
of
1
results
1
Butyl Gallate 98.0+%, TCI America™
CAS: 1083-41-6 Molecular Formula: C11H14O5 Molecular Weight (g/mol): 226.228 MDL Number: MFCD00016432 InChI Key: XOPOEBVTQYAOSV-UHFFFAOYSA-N Synonym: Gallic Acid Butyl Ester PubChem CID: 14128 IUPAC Name: butyl 3,4,5-trihydroxybenzoate SMILES: CCCCOC(=O)C1=CC(=C(C(=C1)O)O)O