accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108321-07-9

IUPAC Name: disodium 2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
Synonyms: disodium 2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
Molecular Weight (g/mol): 406.38
Molecular Formula: C10H20N2Na2O8S2
InChi Key: XMRCLLXGMGEVHS-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC(CN1CCN(CC(O)CS([O-])(=O)=O)CC1)CS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)