CAS RN 108373-05-3
IUPAC Name:
3-bromo-4-ethoxybenzaldehyde
Synonyms:
3-bromo-4-ethoxybenzaldehyde
Molecular Weight (g/mol):
229.07
Molecular Formula:
C9H9BrO2
InChi Key:
TZUUPGZANQRCHD-UHFFFAOYSA-N
SMILES:
CCOC1=C(Br)C=C(C=O)C=C1
1
–
1
of
1
results
1
3-Bromo-4-ethoxybenzaldehyde 97.0+%, TCI America™
CAS: 108373-05-3 Molecular Formula: C9H9BrO2 Molecular Weight (g/mol): 229.073 MDL Number: MFCD00672941 InChI Key: TZUUPGZANQRCHD-UHFFFAOYSA-N PubChem CID: 1494336 IUPAC Name: 3-bromo-4-ethoxybenzaldehyde SMILES: CCOC1=C(C=C(C=C1)C=O)Br