accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOHNSOOClCl

CAS RN 108409-83-2

IUPAC Name: 2,5-dichloro-N-(2-methyl-4-nitrophenyl)benzene-1-sulfonamide
Synonyms: 2,5-dichloro-N-(2-methyl-4-nitrophenyl)benzenesulfonamide
Molecular Weight (g/mol): 361.19
Molecular Formula: C13H10Cl2N2O4S
InChi Key: AXNUEXXEQGQWPA-UHFFFAOYSA-N
SMILES: CC1=C(NS(=O)(=O)C2=C(Cl)C=CC(Cl)=C2)C=CC(=C1)[N+]([O-])=O
Product Line 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results