accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOHNH2HCl

CAS RN 108412-04-0

IUPAC Name: 2-aminopent-4-enoic acid hydrochloride
Synonyms: allylglycine hydrochloride
Molecular Weight (g/mol): 151.59
Molecular Formula: C5H10ClNO2
InChi Key: DIDZZOWASZMNQW-UHFFFAOYNA-N
SMILES: Cl.NC(CC=C)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results