accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1085-12-7

IUPAC Name: heptyl 4-hydroxybenzoate
Synonyms: heptylparaben nipaheptyl p-oxybenzoesaureheptylester staypro WS 7 heptyl P-hydroxybenzoate
Molecular Weight (g/mol): 236.31
Molecular Formula: C14H20O3
InChi Key: ZTJORNVITHUQJA-UHFFFAOYSA-N
SMILES: CCCCCCCOC(=O)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)