accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CSOONSFClCl

CAS RN 1085-98-9

IUPAC Name: ({[dichloro(fluoro)methyl]sulfanyl}(phenyl)sulfamoyl)dimethylamine
Synonyms: dichlofluanid
Molecular Weight (g/mol): 333.22
Molecular Formula: C9H11Cl2FN2O2S2
InChi Key: WURGXGVFSMYFCG-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)N(SC(F)(Cl)Cl)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results