accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 1087-09-8

IUPAC Name: diphenylbut-2-yne-1,4-dione
Synonyms: diphenylbut-2-yne-1,4-dione
Molecular Weight (g/mol): 234.25
Molecular Formula: C16H10O2
InChi Key: BLCJVQFFSUNDMV-UHFFFAOYSA-N
SMILES: O=C(C#CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results