accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1087-21-4

IUPAC Name: 1,3-bis(prop-2-en-1-yl) benzene-1,3-dicarboxylate
Synonyms: 1,3-bis(prop-2-en-1-yl) benzene-1,3-dicarboxylate
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: OOORLLSLMPBSPT-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1=CC(=CC=C1)C(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results