accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH2NCH3

CAS RN 108714-73-4

IUPAC Name: 9,9-dimethyl-9H-fluoren-2-amine
Synonyms: 9,9-dimethylfluoren-2-amine
Molecular Weight (g/mol): 209.29
Molecular Formula: C15H15N
InChi Key: GUTJITRKAMCHSD-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(N)C=C2
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)