accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108906-15-6

IUPAC Name: 1-aminopyrrolidine-2,5-dione hydrochloride
Synonyms: 1-aminopyrrolidine-2,5-dione hydrochloride
Molecular Weight (g/mol): 150.56
Molecular Formula: C4H7ClN2O2
InChi Key: SRKWEKIPNNNHSG-UHFFFAOYSA-N
SMILES: Cl.NN1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results