accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3SiH3CH3CClCl

CAS RN 1089687-03-5

IUPAC Name: dichlorobis(2-ethylhexyl)silane
Synonyms: dichlorobis(2-ethylhexyl)silane
Molecular Weight (g/mol): 325.43
Molecular Formula: C16H34Cl2Si
InChi Key: BXDGSXMFTGZLDT-UHFFFAOYNA-N
SMILES: CCCCC(CC)C[Si](Cl)(Cl)CC(CC)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results