accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH

CAS RN 109-01-3

IUPAC Name: 1-methylpiperazine
Synonyms: piperazine, 1-methyl-
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: PVOAHINGSUIXLS-UHFFFAOYSA-N
SMILES: CN1CCNCC1
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (8)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (7)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (3)
  • (7)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (8)
  • (6)
  • (3)
  • (3)
  • (1)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (7)
  • (1)
  • (1)