accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3

CAS RN 109-19-3

IUPAC Name: butyl 3-methylbutanoate
Synonyms: butyl isovalerate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: AYWJSCLAAPJZEF-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results