accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 109-29-5

IUPAC Name: 1-oxacycloheptadecan-2-one
Synonyms: hexadecanolactone
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: LOKPJYNMYCVCRM-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCCCCCO1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)