accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109-53-5

IUPAC Name: 1-(ethenyloxy)-2-methylpropane
Synonyms: isobutyl vinyl ether
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: OZCMOJQQLBXBKI-UHFFFAOYSA-N
SMILES: CC(C)COC=C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)