CAS RN 109-61-5
CAS RN 109-61-5
IUPAC Name:
propyl carbonochloridate
Synonyms:
carbonochloridic acid, propyl ester
Molecular Weight (g/mol):
122.55
Molecular Formula:
C4H7ClO2
InChi Key:
QQKDTTWZXHEGAQ-UHFFFAOYSA-N
SMILES:
CCCOC(Cl)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Propyl chloroformate, 98%
CAS: 109-61-5 Molecular Weight (g/mol): 122.55 InChI Key: QQKDTTWZXHEGAQ-UHFFFAOYSA-N Synonym: propyl chloroformate,propyl chlorocarbonate,n-propyl chloroformate,carbonochloridic acid, propyl ester,chloroformicacidpropylester,formic acid, chloro-, propyl ester,unii-a5y2t003ju,n-propylchloroformate,propylchloroformate,chloroformic acid propyl ester PubChem CID: 7998 IUPAC Name: propyl carbonochloridate