accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 109-64-8

IUPAC Name: 1,3-dibromopropane
Synonyms: 1,3-dibromopropane
Molecular Weight (g/mol): 201.89
Molecular Formula: C3H6Br2
InChi Key: VEFLKXRACNJHOV-UHFFFAOYSA-N
SMILES: BrCCCBr
Molecular Weight (g/mol) 
  • (10)
  • (2)
Boiling Point 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (3)
  • (6)
  • (10)
  • (2)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (9)
  • (1)